2-Bromo-6-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

B1878-25G B1878-5G
TCB1878-25GEA 224.83 CAD
TCB1878-25G TCB1878-5G
2-Bromo-6-nitrotoluene ≥98.0% (by GC)
2-Bromo-6-nitrotoluene

CAS Number: 55289-35-5
MDL Number: MFCD00009792
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 40
Flash Point (°C): 110

Formula: C₇H₆BrNO₂
Boiling Pt: 136 °C
Melting Pt: 40 °C
Flash Pt: 110 °C
MDL Number: MFCD00009792
CAS Number: 55289-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=325fbd3c95dbc684947228eccd5e7fad56b3fe4b&vid=c697187f6cd5ebd1aa329c7186821d62bd704b4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR