2-Bromo-3,3,3-trifluoropropene ≥97.0% (by GC)
Supplier: TCI America
B1801-25G
B1801-5G
TCB1801-25GEA
325.48
CAD
TCB1801-25G
TCB1801-5G
2-Bromo-3,3,3-trifluoropropene ≥97.0% (by GC)
2-Bromo-3,3,3-trifluoropropene
CAS Number: 1514-82-5
MDL Number: MFCD00077469
Molecular Formula: C3H2BrF3
Molecular Weight: 174.95
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 34
Specific Gravity (20/20): 1.66
Storage Temperature: 0-10°C
Formula:
C₃H₂BrF₃ Boiling Pt: 34 °C Density: 1.66 (20 °C) |
MDL Number:
MFCD00077469 CAS Number: 1514-82-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...