1-Bromo-3,3-dimethylbutanone ≥95.0% (by GC)
Supplier: TCI America
B1490-25G
B1490-5G
TCB1490-25GEA
447.93
CAD
TCB1490-25G
TCB1490-5G
1-Bromo-3,3-dimethylbutanone ≥95.0% (by GC)
1-Bromo-3,3-dimethylbutanone
CAS Number: 5469-26-1
MDL Number: MFCD00000206
Molecular Formula: C6H11BrO
Molecular Weight: 179.06
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Flash Point (°C): 66
Specific Gravity (20/20): 1.33
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Bromomethyl tert-Butyl Ketone, 1-Bromopinacolin, 1-Bromopinacolone
Formula:
C₆H₁₁BrO Boiling Pt: 188 °C Density: 1.33 (20 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00000206 CAS Number: 5469-26-1 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...