4-Bromoaniline ≥99.0% (by GC)

Supplier: TCI America

B1393-25G B1393-500G B1393-100G
TCB1393-25GEA 79.66 CAD
TCB1393-25G TCB1393-500G TCB1393-100G
4-Bromoaniline ≥99.0% (by GC)
4-Bromoaniline

CAS Number: 106-40-1
MDL Number: MFCD00007822
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 65

Formula: C₆H₆BrN
Boiling Pt: 124 °C
Melting Pt: 65 °C
MDL Number: MFCD00007822
CAS Number: 106-40-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01c380f8aaff047017260221a2427168fc68358d&vid=021e60b67c6a99df117f7b0c39f33a1f0d987a0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR