3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Dienestrol Diacetate

B1366-100MG B1366-1G
TCB1366-100MGEA 62.88 CAD
TCB1366-100MG TCB1366-1G
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene ≥95.0% (by GC)
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene

CAS Number: 84-19-5
MDL Number: MFCD00063286
Molecular Formula: C22H22O4
Molecular Weight: 350.41
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 122


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Dienestrol Diacetate

Formula: C₂₂H₂₂O₄
MW: 350.41 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00063286
CAS Number: 84-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11adb9ec2f7d84140f0c249d1a5215fcdbaa01dd&vid=11adb9ec2f7d84140f0c249d1a5215fcdbaa01dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR