1,4-Butanediol diglycidyl ether ≥93.0% (by GC)

Supplier: TCI America

Synonyms: Araldite RD-2, 1,4-Bis(oxiran-2-ylmethoxy)butane

B0964-250ML B0964-25ML
TCB0964-250MLEA 1211.86 CAD
TCB0964-250ML TCB0964-25ML
1,4-Butanediol diglycidyl ether ≥93.0% (by GC)
1,4-Butanediol diglycidyl ether

CAS Number: 2425-79-8
MDL Number: MFCD00005146
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Melting point (°C): -21
Flash Point (°C): 140
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers
Other Product Names / Synonyms: 1,4-Bis(2,3-epoxypropoxy)butane, 1,4-Diglycidyloxybutane, Tetramethylene Glycol Diglycidyl Ether

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 160 °C
Melting Pt: –21 °C
Density: 1.07 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00005146
CAS Number: 2425-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e216f098f41b6ed837cbe197f15952824f7c3a84&vid=e216f098f41b6ed837cbe197f15952824f7c3a84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR