2'-Bromoacetophenone ≥98.0%

Supplier: TCI America

Synonyms: o-Bromoacetophenone

B0538-25G B0538-5G
TCB0538-25GEA 205.4 CAD
TCB0538-25G TCB0538-5G
2'-Bromoacetophenone ≥98.0%
2'-Bromoacetophenone

CAS Number: 2142-69-0
MDL Number: MFCD00000067
Molecular Formula: C8H7BrO
Molecular Weight: 199.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 110
Specific Gravity (20/20): 1.49

Formula: C₈H₇BrO
Boiling Pt: 136 °C
Density: 1.49 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000067
CAS Number: 2142-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3c55d7b46629598acd5f108a1b1336941a8c82e&vid=e3c55d7b46629598acd5f108a1b1336941a8c82e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR