4-Butylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0241-25G B0241-5G
TCB0241-25GEA 173.8 CAD
TCB0241-25G TCB0241-5G
4-Butylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Butylbenzoic acid

CAS Number: 20651-71-2
MDL Number: MFCD00002571
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 100

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00002571
CAS Number: 20651-71-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1133edbc36bf96dec1bbfaa3baa3600d5e0498d8&vid=b6f278e273a0831549c55347e4dab8b4c3cd10a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR