2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-((2-(Hydroxymethyl)phenyl)methylene)diphenol, Phenolphthalol

B0100-25G B0100-5G
TCB0100-25GEA 811.65 CAD
TCB0100-25G TCB0100-5G
2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol ≥98.0%
2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol

CAS Number: 81-92-5
MDL Number: MFCD00059144
Molecular Formula: C20H18O3
Molecular Weight: 306.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 204


Other Product Names / Synonyms: Phenolphthalol

Formula: C₂₀H₁₈O₃
MW: 306.36 g/mol
Melting Pt: 204 °C
MDL Number: MFCD00059144
CAS Number: 81-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee914e8514b9f7344ad3161cd22e1afe203ded92&vid=ee914e8514b9f7344ad3161cd22e1afe203ded92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR