2,3-Benzofluorene ≥97.0% (by GC)

Supplier: TCI America

B0059-100MG B0059-1G
TCB0059-100MGEA 127.98 CAD
TCB0059-100MG TCB0059-1G
2,3-Benzofluorene ≥97.0% (by GC)
2,3-Benzofluorene

CAS Number: 243-17-4
MDL Number: MFCD00003595
Molecular Formula: C17H12
Molecular Weight: 216.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 402
Melting point (°C): 213


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2,3-Benzfluorene

Formula: C₁₇H₁₂
Boiling Pt: 402 °C
Melting Pt: 213 °C
MDL Number: MFCD00003595
CAS Number: 243-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da9d4f9daf16ffed7f4f33dcb0cdc3e5b69080ed&vid=da9d4f9daf16ffed7f4f33dcb0cdc3e5b69080ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR