4-Amino-2-chloro-5-fluoropyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2809-1G A2809-5G
TCA2809-1GEA 99.54 CAD
TCA2809-1G TCA2809-5G
4-Amino-2-chloro-5-fluoropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2-chloro-5-fluoropyrimidine

CAS Number: 155-10-2
MDL Number: MFCD00057344
Molecular Formula: C4H3ClFN3
Molecular Weight: 147.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 195

Formula: C₄H₃ClFN₃
Melting Pt: 195 °C
MDL Number: MFCD00057344
CAS Number: 155-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1de92d9f99a1d3545482b549e0b03f44a80a9a02&vid=1de92d9f99a1d3545482b549e0b03f44a80a9a02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR