2-Acetyl-6-methoxypyridine ≥97.0%

Supplier: TCI America

Synonyms: 1-(6-Methoxy-2-pyridinyl)ethanone

A2802-1G A2802-5G
TCA2802-1GEA 232.26 CAD
TCA2802-1G TCA2802-5G
2-Acetyl-6-methoxypyridine ≥97.0%
2-Acetyl-6-methoxypyridine

CAS Number: 21190-93-2
MDL Number: MFCD08276836
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 41
Flash Point (°C): 94

Formula: C₈H₉NO₂
Boiling Pt: 85 °C
Melting Pt: 41 °C
Flash Pt: 94 °C
MDL Number: MFCD08276836
CAS Number: 21190-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba56c556f30300f8ddcaff30b6a317daf9b8617c&vid=ba56c556f30300f8ddcaff30b6a317daf9b8617c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR