2-Amino-5-chloropyrazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2790-1G
A2790-5G
TCA2790-1GEA
79
CAD
TCA2790-1G
TCA2790-5G
2-Amino-5-chloropyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-chloropyrazine
CAS Number: 33332-29-5
MDL Number: MFCD03423596
Molecular Formula: C4H4ClN3
Molecular Weight: 129.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Formula:
C₄H₄ClN₃ Melting Pt: 134 °C |
MDL Number:
MFCD03423596 CAS Number: 33332-29-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...