1,3-Benzoxazol-2-amine ≥98.0%

Supplier: TCI America

Synonyms: Benzo[d]oxazol-2-amine, 2-Aminobenzoxazole

A2774-25G A2774-5G
TCA2774-25GEA 1336.68 CAD
TCA2774-25G TCA2774-5G
1,3-Benzoxazol-2-amine ≥98.0%
1,3-Benzoxazol-2-amine

CAS Number: 4570-41-6
MDL Number: MFCD00182850
Molecular Formula: C7H6N2O
Molecular Weight: 134.14
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 130

Formula: C₇H₆N₂O
Melting Pt: 130 °C
MDL Number: MFCD00182850
CAS Number: 4570-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0ea1af912a3f520e8ea8321fef04675ab27e6a6&vid=e0ea1af912a3f520e8ea8321fef04675ab27e6a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR