L-Alanine benzyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-Ala-OBzl.HCl, H-Ala-OBzl·HCl
A2771-25G
A2771-5G
TCA2771-25GEA
225.94
CAD
TCA2771-25G
TCA2771-5G
L-Alanine benzyl ester hydrochloride ≥98.0%
L-Alanine benzyl ester hydrochloride
CAS Number: 5557-83-5
MDL Number: MFCD00054340
Molecular Formula: C10H13NO2
Molecular Weight: 215.68
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 138
Specific rotation [a]20/D: -10 deg (C=1, 0.1mol/L HCl)
Storage Temperature: <0°C
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Ala-OBzl.HCl
Formula:
C₁₀H₁₃NO₂ MW: 215.68 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD00054340 CAS Number: 5557-83-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...