4'-Acetamido-2'-methylacetophenone ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Acetyl-3-methylphenyl)acetamide

A2767-25G A2767-5G
TCA2767-25GEA 157.05 CAD
TCA2767-25G TCA2767-5G
4'-Acetamido-2'-methylacetophenone ≥98.0%
4'-Acetamido-2'-methylacetophenone

CAS Number: 34956-31-5
MDL Number: MFCD02258876
Molecular Formula: C11H13NO2
Molecular Weight: 191.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139


Other Product Names / Synonyms: N-(4-Acetyl-3-methylphenyl)acetamide

Formula: C₁₁H₁₃NO₂
Melting Pt: 139 °C
MDL Number: MFCD02258876
CAS Number: 34956-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d20b414a0603d2787af37ea879f2bec4d35574a2&vid=d20b414a0603d2787af37ea879f2bec4d35574a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR