2-Amino-4'-fluorobenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (2-Aminophenyl)(4-fluorophenyl)methanone

A2760-1G A2760-5G
TCA2760-1GEA 33.18 CAD
TCA2760-1G TCA2760-5G
2-Amino-4'-fluorobenzophenone ≥98.0%
2-Amino-4'-fluorobenzophenone

CAS Number: 3800-06-4
Molecular Formula: C13H10FNO
Molecular Weight: 215.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 129

Formula: C₁₃H₁₀FNO
Melting Pt: 129 °C
MDL Number: MFCD06658166
CAS Number: 3800-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f05ff7cf35296d298509b04bcab5b99950f89438&vid=f05ff7cf35296d298509b04bcab5b99950f89438&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR