5-Amino-2-(4-aminophenyl)benzimidazole ≥98.0% (by HPLC)
Supplier: TCI America
A2759-1G
A2759-5G
TCA2759-1GEA
71.1
CAD
TCA2759-1G
TCA2759-5G
5-Amino-2-(4-aminophenyl)benzimidazole ≥98.0% (by HPLC)
5-Amino-2-(4-aminophenyl)benzimidazole
CAS Number: 7621-86-5
MDL Number: MFCD00451475
Molecular Formula: C13H12N4
Molecular Weight: 224.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 236
Lambda max.: 335 nm (EtOH)
Formula:
C₁₃H₁₂N₄ Melting Pt: 236 °C |
MDL Number:
MFCD00451475 CAS Number: 7621-86-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...