6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 6-Acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one

A2755-25G A2755-5G
TCA2755-25GEA 276.03 CAD
TCA2755-25G TCA2755-5G
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by HPLC, total nitrogen)
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one

CAS Number: 26518-71-8
MDL Number: MFCD01321313
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 196

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 196 °C
MDL Number: MFCD01321313
CAS Number: 26518-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a9ac8234b2c745d99d5caa2e9105c12b98cdd38&vid=3a9ac8234b2c745d99d5caa2e9105c12b98cdd38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR