3-Amino-4-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-Amino-4-methylbenzeneboronic Acid, 3-Amino-p-tolylboronic Acid, 3-Amino-4-methylphenylboronic Acid

A2745-1G A2745-5G
TCA2745-1GEA 202.24 CAD
TCA2745-1G TCA2745-5G
3-Amino-4-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Amino-4-methylphenylboronic acid

CAS Number: 22237-12-3
MDL Number: MFCD01074640
Molecular Formula: C7H10BNO2
Molecular Weight: 150.97
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: 3-Amino-4-methylbenzeneboronic Acid, 3-Amino-p-tolylboronic Acid

Formula: C₇H₁₀BNO₂
MW: 150.97 g/mol
MDL Number: MFCD01074640
CAS Number: 22237-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b10225343a8469eb425fc3852a1162103b38556&vid=9b10225343a8469eb425fc3852a1162103b38556&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR