Arabinofuranosyl-guanine hydrate ≥98.0% (by HPLC)

Supplier: TCI America

A2739-10MG A2739-50MG
TCA2739-10MGEA 88.48 CAD
TCA2739-10MG TCA2739-50MG
Arabinofuranosyl-guanine hydrate ≥98.0% (by HPLC)
Arabinofuranosyl-guanine hydrate

CAS Number: 38819-10-2
MDL Number: MFCD00065486
Molecular Formula: C10H13N5O5
Molecular Weight: 283.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 225
Lambda max.: 252 nmStorage Temperature: 0-10°C


Product Categories: Bioscience, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: Ara-G, Guanine 9-beta-D-Arabinofuranoside

Formula: C₁₀H₁₃N₅O₅
Melting Pt: 225 °C
MDL Number: MFCD00065486
CAS Number: 38819-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d058be3b190e38c73c389e4a4bc6cd5ae91683d&vid=3d058be3b190e38c73c389e4a4bc6cd5ae91683d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR