4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC)

Supplier: TCI America

A2737-1G A2737-200MG
TCA2737-1GEA 178.54 CAD
TCA2737-1G TCA2737-200MG
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC)
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine

CAS Number: 5334-30-5
MDL Number: MFCD00022901
Molecular Formula: C11H9N5
Molecular Weight: 211.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 216
Lambda max.: 240 nm (H2O (pH 1))Storage Temperature: >-20°C


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: PP 3

Formula: C₁₁H₉N₅
Melting Pt: 216 °C
MDL Number: MFCD00022901
CAS Number: 5334-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=978079615413aad1aa6ec42be9e4f640ce4bc274&vid=978079615413aad1aa6ec42be9e4f640ce4bc274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR