3-Acetyl-2,4-dimethylpyrrole ≥98.0% (by GC)

Supplier: TCI America

A2734-1G A2734-5G
TCA2734-1GEA 97.96 CAD
TCA2734-1G TCA2734-5G
3-Acetyl-2,4-dimethylpyrrole ≥98.0% (by GC)
3-Acetyl-2,4-dimethylpyrrole

CAS Number: 2386-25-6
MDL Number: MFCD00005221
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 173
Melting point (°C): 140
Lambda max.: 282 nm (EtOH)

Formula: C₈H₁₁NO
Boiling Pt: 173 °C
Melting Pt: 140 °C
MDL Number: MFCD00005221
CAS Number: 2386-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=767f583ac5764e8462112a63bd23613ea257ab72&vid=767f583ac5764e8462112a63bd23613ea257ab72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR