25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90.0% (by HPLC)

Supplier: TCI America

A2727-100MG A2727-25MG
TCA2727-100MGEA 213.3 CAD
TCA2727-100MG TCA2727-25MG
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90.0% (by HPLC)
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane

CAS Number: 869718-80-9
MDL Number: MFCD13184962
Molecular Formula: C17H35N3O8
Molecular Weight: 409.48
Purity/Analysis Method: >90.0% (HPLC)
Form: Clear Liquid
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, PEGylation Reagents & PEG Spacers, PEGylation Reagents
Other Product Names / Synonyms: O-(2-Azidoethyl)-O'-methylheptaethylene Glycol, mPEG8-Azide, mPEG8-N3, Methyl-PEG8-Azide

Formula: C₁₇H₃₅N₃O₈
MDL Number: MFCD13184962
CAS Number: 869718-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b2b690b4098ebc08d1f1629f6e265566829ecc8&vid=59178173258cc593bb41d4efbe326e318feff689&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR