4-(Methylsulfonyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2725-25G A2725-100G
TCA2725-25GEA 82.16 CAD
TCA2725-25G TCA2725-100G
4-(Methylsulfonyl)aniline ≥98.0% (by GC, titration analysis)
4-(Methylsulphonyl)aniline

CAS Number: 5470-49-5
MDL Number: MFCD00025363
Molecular Formula: C7H9NO2S
Molecular Weight: 171.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 4-Aminophenyl Methyl Sulfone

Formula: C₇H₉NO₂S
MW: 171.21 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00025353
CAS Number: 5470-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59adfcb65e6af76c1460bee059e5a52fa339d6d&vid=d59adfcb65e6af76c1460bee059e5a52fa339d6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR