3-Amino-N-(tert-butoxycarbonyl)benzylamine 98,0
Supplier: TCI America
Synonyms:
alpha-(Boc-amino)-m-toluidine, 3-Amino-N-Boc-benzylamine, tert-Butyl (3-Aminobenzyl)carbamate, alpha-(tert-Butoxycarbonyl)amino-m-toluidine, 3-[(tert-Butoxycarbonylamino)methyl]aniline, 3-[(Boc-amino)methyl]aniline, (3-Aminobenzyl)carbamic Acid tert-Butyl, tert-Butyl 3-aminobenzylcarbamate
A2718-1G
A2718-5G
TCA2718-1GEA
478.74
CAD
TCA2718-1G
TCA2718-5G
3-Amino-N-(tert-butoxycarbonyl)benzylamine 98,0
3-Amino-N-(tert-butoxycarbonyl)benzylamine
CAS Number: 147291-66-5
MDL Number: MFCD04974010
Molecular Formula: C12H18N2O2
Molecular Weight: 222.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 50
Formula:
C₁₂H₁₈N₂O₂ MW: 222.29 g/mol Melting Pt: 50 °C |
MDL Number:
MFCD04974010 CAS Number: 147291-66-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...