6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2717-1G A2717-5G
TCA2717-1GEA 54.51 CAD
TCA2717-1G TCA2717-5G
6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
6-Acetoxy-7-methoxy-3H-quinazolin-4-one

CAS Number: 179688-53-0
MDL Number: MFCD09951978
Molecular Formula: C11H10N2O4
Molecular Weight: 234.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 297


Other Product Names / Synonyms: 6-Acetoxy-7-methoxy-4(3H)-quinazolinone, 6-Acetoxy-7-methoxy-3,4-dihydroquinazolin-4-one, 6-Acetoxy-7-methoxy-4-hydroxyquinazoline, 6-Acetoxy-7-methoxy-4-quinazolinol

Formula: C₁₁H₁₀N₂O₄
Melting Pt: 297 °C
MDL Number: MFCD09951978
CAS Number: 179688-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e167b04a52bb1eb0242c1581002d331707630ce&vid=403a48cc10acaac00605dc6d7bb948b56919c579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR