5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98.0%

Supplier: TCI America

Synonyms: 4,6-Dichloro-2-(propylthio)pyrimidin-5-amine

A2716-1G A2716-5G
TCA2716-1GEA 93.22 CAD
TCA2716-1G TCA2716-5G
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98.0%
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine

CAS Number: 145783-15-9
MDL Number: MFCD16875503
Molecular Formula: C7H9Cl2N3S
Molecular Weight: 238.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₇H₉Cl₂N₃S
MW: 238.13 g/mol
MDL Number: MFCD16875503
CAS Number: 145783-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d90f15c23789995eae5e2c5a316808770d720fc2&vid=d90f15c23789995eae5e2c5a316808770d720fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR