tert-Butyl-6-aminopyridin-2-ylcarbamate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A2708-1G A2708-5G
TCA2708-1GEA 155.31 CAD
TCA2708-1G TCA2708-5G
tert-Butyl-6-aminopyridin-2-ylcarbamate ≥97.0% (by titrimetric analysis)
tert-Butyl-6-aminopyridin-2-ylcarbamate

CAS Number: 322690-31-3
MDL Number: MFCD09037865
Molecular Formula: C10H15N3O2
Molecular Weight: 209.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 127


Other Product Names / Synonyms: 2-Amino-6-(Boc-amino)pyridine

Formula: C₁₀H₁₅N₃O₂
Melting Pt: 127 °C
MDL Number: MFCD09037865
CAS Number: 322690-31-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd2e1b63ff0c2233d64f7d0c35ab384401d31ffe&vid=bd2e1b63ff0c2233d64f7d0c35ab384401d31ffe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR