2-Acetamido-6-aminopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-(6-Aminopyridin-2-yl)acetamide

A2707-25G A2707-5G
TCA2707-25GEA 496.12 CAD
TCA2707-25G TCA2707-5G
2-Acetamido-6-aminopyridine ≥98.0% (by GC, titration analysis)
2-Acetamido-6-aminopyridine

CAS Number: 1075-62-3
MDL Number: MFCD01099052
Molecular Formula: C7H9N3O
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160

Formula: C₇H₉N₃O
MW: 151.17 g/mol
Melting Pt: 160 °C
MDL Number: MFCD01099052
CAS Number: 1075-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46f5e8c44ff17d06f6b2096077b9497e5ce6c1b&vid=c46f5e8c44ff17d06f6b2096077b9497e5ce6c1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR