5'-Acetamido-2'-hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(3-Acetyl-4-hydroxyphenyl)acetamide

A2698-1G A2698-5G
TCA2698-1GEA 89.27 CAD
TCA2698-1G TCA2698-5G
5'-Acetamido-2'-hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
5'-Acetamido-2'-hydroxyacetophenone

CAS Number: 7298-67-1
MDL Number: MFCD00100490
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 168


Other Product Names / Synonyms: 3'-Acetyl-4'-hydroxyacetanilide

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00100490
CAS Number: 7298-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3af0badc3b67161272516f4b897aa20f12532303&vid=3af0badc3b67161272516f4b897aa20f12532303&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR