2-Amino-4-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2690-1G A2690-5G
TCA2690-1GEA 75.84 CAD
TCA2690-1G TCA2690-5G
2-Amino-4-methoxypyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-methoxypyridine

CAS Number: 10201-73-7
MDL Number: MFCD07437849
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118
Lambda max.: 280 nm (CHCl3)

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 118 °C
MDL Number: MFCD07437849
CAS Number: 10201-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddfb056ede954f68f70d17d6fcb1e99fb2cf5910&vid=ddfb056ede954f68f70d17d6fcb1e99fb2cf5910&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR