3-Amino-4-bromo-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

A2688-1G A2688-5G
TCA2688-1GEA 48.98 CAD
TCA2688-1G TCA2688-5G
3-Amino-4-bromo-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-4-bromo-1H-pyrazole

CAS Number: 16461-94-2
MDL Number: MFCD00082728
Molecular Formula: C3H4BrN3
Molecular Weight: 161.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₃H₄BrN₃
MW: 161.99 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00082728
CAS Number: 16461-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8412ac22d8906d7d80c056ea2a92112fb7cd086e&vid=8412ac22d8906d7d80c056ea2a92112fb7cd086e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR