N-Acetyl-DL-2-phenylglycine 98,0

Supplier: TCI America

Synonyms: N-Acetyl-DL-phenylglycine, Ac-DL-Phg-OH, (+/-)-alpha-(Acetylamino)phenylacetic Acid

A2687-25G A2687-5G
TCA2687-25GEA 167.48 CAD
TCA2687-25G TCA2687-5G
N-Acetyl-DL-2-phenylglycine 98,0
N-Acetyl-DL-2-phenylglycine

CAS Number: 15962-46-6
MDL Number: MFCD00055924
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (+/-)-alpha-(Acetylamino)phenylacetic Acid, Ac-DL-Phg-OH

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00055924
CAS Number: 15962-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f299fc44f5affa36ec09806517b67a0f0fe4ea19&vid=f299fc44f5affa36ec09806517b67a0f0fe4ea19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR