1,4-Anthraquinone ≥95.0%
Supplier: TCI America
Synonyms:
Anthracene-1,4-dione
A2686-1G
A2686-5G
TCA2686-1GEA
153.99
CAD
TCA2686-1G
76218-748
1,4-Anthraquinone ≥95.0%
1,4-Anthraquinone
CAS Number: 635-12-1
MDL Number: MFCD00013724
Molecular Formula: C14H8O2
Molecular Weight: 208.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 218
Lambda max.: 298 nm (EtOH)
Other Product Names / Synonyms: 1,4-Anthracenedione
Formula:
C₁₄H₈O₂ Melting Pt: 218 °C |
MDL Number:
MFCD00013724 CAS Number: 635-12-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...