≥98.0%

Supplier: TCI America

Synonyms: Phenylacetic acid Pentyl ester, Phenylacetic acid Amyl ester, Amyl Phenylacetate, Pentyl Phenylacetate

A2680-25ML A2680-5ML
TCA2680-25MLEA 289.3 CAD
TCA2680-25ML TCA2680-5ML
≥98.0%
Pentyl phenylacetate

CAS Number: 5137-52-0
MDL Number: MFCD00027272
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 269
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Phenylacetic Acid Amyl Ester, Pentyl Phenylacetate, Phenylacetic Acid Pentyl Ester

Formula: C₁₃H₁₈O₂
Boiling Pt: 269 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00027272
CAS Number: 5137-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0ccd59f341355166ceb4fc15895b07f430c4b69&vid=a0ccd59f341355166ceb4fc15895b07f430c4b69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR