2-Amino-5-(benzylthio)-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-(Benzylthio)-1,3,4-thiadiazol-2-amine, 5-(Benzylthio)-1,3,4-thiadiazole-2-amine

A2677-1G A2677-5G
TCA2677-1GEA 268.6 CAD
TCA2677-1G TCA2677-5G
2-Amino-5-(benzylthio)-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-(benzylthio)-1,3,4-thiadiazole

CAS Number: 25660-71-3
MDL Number: MFCD00184237
Molecular Formula: C9H9N3S2
Molecular Weight: 223.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 5-(Benzylthio)-1,3,4-thiadiazole-2-amine

Formula: C₉H₉N₃S₂
MW: 223.31 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00184237
CAS Number: 25660-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89bc18369eb959e73349bd9b046b47935ff796e3&vid=89bc18369eb959e73349bd9b046b47935ff796e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR