2-Amino-5-bromo-3-pyridinol ≥98.0% (by HPLC)
Supplier: TCI America
A2673-1G
A2673-5G
TCA2673-1GEA
54.51
CAD
TCA2673-1G
TCA2673-5G
2-Amino-5-bromo-3-pyridinol ≥98.0% (by HPLC)
2-Amino-5-bromo-3-pyridinol
CAS Number: 39903-01-0
MDL Number: MFCD09744143
Molecular Formula: C5H5BrN2O
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 200
Other Product Names / Synonyms: 2-Amino-5-bromo-3-pyridinol
Formula:
C₅H₅BrN₂O Melting Pt: 200 °C |
MDL Number:
MFCD09744143 CAS Number: 39903-01-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...