2-(4-Amino-2-nitroanilino)ethanol ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

A2671-25G A2671-5G
TCA2671-25GEA 341.28 CAD
TCA2671-25G TCA2671-5G
2-(4-Amino-2-nitroanilino)ethanol ≥95.0% (by HPLC, titration analysis)
2-(4-Amino-2-nitroanilino)ethanol

CAS Number: 2871-01-4
MDL Number: MFCD00071801
Molecular Formula: C8H11N3O3
Molecular Weight: 197.19
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 126


Other Product Names / Synonyms: 4-Amino-N-(2-hydroxyethyl)-2-nitroaniline, N1-(2-Hydroxyethyl)-2-nitro-1,4-phenylenediamine

Formula: C₈H₁₁N₃O₃
Melting Pt: 126 °C
MDL Number: MFCD00071801
CAS Number: 2871-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cefda17bd3556e874acafa11f11f8ebf5916244&vid=8cefda17bd3556e874acafa11f11f8ebf5916244&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR