3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetyl-5-chlorothiophene-2-sulfonamide, 3-Acetyl-5-chloro-2-thiophenesulphonamide, 3-Acetyl-5-chlorothiophene-2-sulphonamide, 3-Acetyl-5-chloro-2-thiophenesulfonamide

A2667-1G A2667-5G
TCA2667-1GEA 135.88 CAD
TCA2667-1G TCA2667-5G
3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene ≥98.0%
3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene

CAS Number: 160982-10-5
MDL Number: MFCD09033309
Molecular Formula: C6H6ClNO3S2
Molecular Weight: 239.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 3-Acetyl-5-chloro-2-thiophenesulfonamide

Formula: C₆H₆ClNO₃S₂
Melting Pt: 182 °C
MDL Number: MFCD09033309
CAS Number: 160982-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16128774cc627ddcef95c30115bbaf01c760fbfe&vid=16128774cc627ddcef95c30115bbaf01c760fbfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR