2-Amino-4-(p-tolyl)thiazole ≥98.0%

Supplier: TCI America

Synonyms: 4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine, 4-(p-Tolyl)thiazol-2-amine, 2-Amino-4-(4-methylphenyl)thiazole

A2666-25G A2666-5G
TCA2666-25GEA 409.22 CAD
TCA2666-25G TCA2666-5G
2-Amino-4-(p-tolyl)thiazole ≥98.0%
2-Amino-4-(p-tolyl)thiazole

CAS Number: 2103-91-5
MDL Number: MFCD00170264
Molecular Formula: C10H10N2S
Molecular Weight: 190.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137
Lambda max.: 266 nm (EtOH)


Other Product Names / Synonyms: 2-Amino-4-(4-methylphenyl)thiazole, 4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine

Formula: C₁₀H₁₀N₂S
MW: 190.26 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00170264
CAS Number: 2103-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b56b4764d4cca226e0d3a86d52c62dab73905441&vid=b56b4764d4cca226e0d3a86d52c62dab73905441&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR