N-Allyl-p-toluenesulfonamide ≥98.0% (by GC)

Supplier: TCI America

A2665-1G A2665-5G
TCA2665-1GEA 175.22 CAD
TCA2665-1G TCA2665-5G
N-Allyl-p-toluenesulfonamide ≥98.0% (by GC)
N-Allyl-p-toluenesulfonamide

CAS Number: 50487-71-3
MDL Number: MFCD00182633
Molecular Formula: C10H13NO2S
Molecular Weight: 211.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63

Formula: C₁₀H₁₃NO₂S
Melting Pt: 63 °C
MDL Number: MFCD00182633
CAS Number: 50487-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7f45010167ee7f316389ad387d7462f4064f895&vid=a7f45010167ee7f316389ad387d7462f4064f895&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR