Anthracene-9,10-dicarboxaldehyde ≥98.0% (by GC)
Supplier: TCI America
A2664-1G
A2664-5G
TCA2664-1GEA
237
CAD
TCA2664-1G
TCA2664-5G
Anthracene-9,10-dicarboxaldehyde ≥98.0% (by GC)
Anthracene-9,10-dicarboxaldehyde
CAS Number: 7044-91-9
MDL Number: MFCD00045386
Molecular Formula: C16H10O2
Molecular Weight: 234.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 244
Lambda max.: 430 nm (Diphenyl ether)
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9,10-Diformylanthracene
Formula:
C₁₆H₁₀O₂ Melting Pt: 244 °C |
MDL Number:
MFCD00045386 CAS Number: 7044-91-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...