DL-2-Allylglycine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: DL-2-Amino-4-pentenoic acid, DL-2-Allylglycine, 2-Amino-4-pentenoic Acid, 2-Aminopent-4-enoic acid

A2663-1G A2663-5G
TCA2663-1GEA 173.8 CAD
TCA2663-1G TCA2663-5G
DL-2-Allylglycine ≥98.0% (by HPLC)
DL-2-Allylglycine

CAS Number: 7685-44-1
MDL Number: MFCD00063103
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 220
Storage Temperature: 0-10°C


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-4-pentenoic Acid

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00063103
CAS Number: 7685-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e8e1fb31ae4a2d6a1453a3cfbe2db4bde2f11aa&vid=3e8e1fb31ae4a2d6a1453a3cfbe2db4bde2f11aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR