N-Acetyl-3,5-dimethyl-1-adamantanamine ≥97.0%
Supplier: TCI America
Synonyms:
N-Acetylmemantine, 1-Acetamido-3,5-dimethyladamantane, 1-Actamido-3,5-dimethyladmantane, N-(3,5-Dimethyladamantan-1-yl)acetamide
A2653-1G
A2653-5G
TCA2653-1GEA
330.22
CAD
TCA2653-1G
TCA2653-5G
N-Acetyl-3,5-dimethyl-1-adamantanamine ≥97.0%
N-Acetyl-3,5-dimethyl-1-adamantanamine
CAS Number: 19982-07-1
MDL Number: MFCD06656139
Molecular Formula: C14H23NO
Molecular Weight: 221.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 112
Other Product Names / Synonyms: 1-Acetamido-3,5-dimethyladamantane, N-Acetylmemantine
Formula:
C₁₄H₂₃NO MW: 221.34 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD06656139 CAS Number: 19982-07-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...