Acetamidoxime ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Acetamide oxime

A2646-25G A2646-5G
TCA2646-25GEA 415.54 CAD
TCA2646-25G TCA2646-5G
Acetamidoxime ≥97.0% (by GC, titration analysis)
Acetamidoxime

CAS Number: 22059-22-9
MDL Number: MFCD00603514
Molecular Formula: C2H6N2O
Molecular Weight: 74.08
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: Acetamide Oxime

Formula: C₂H₆N₂O
Melting Pt: 138 °C
MDL Number: MFCD00603514
CAS Number: 22059-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28f82f1ebb37e2cfde585c37d0e52e4477e0511a&vid=de30d743010361c03a1a36bf6f9bd20d3b76eb79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR