4-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Iodopyridin-4-amine, 3-Iodo-4-pyridylamine
A2641-1G
A2641-5G
TCA2641-1GEA
104.12
CAD
TCA2641-1G
TCA2641-5G
4-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-iodopyridine
CAS Number: 88511-27-7
MDL Number: MFCD02663853
Molecular Formula: C5H5IN2
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 98
Formula:
C₅H₅IN₂ MW: 220.01 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD02663853 CAS Number: 88511-27-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...