2-Amino-9,9-dimethylfluorene ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

A2634-1G A2634-200MG
TCA2634-1GEA 102.7 CAD
TCA2634-1G TCA2634-200MG
2-Amino-9,9-dimethylfluorene ≥99.0% (by HPLC, titration analysis)
2-Amino-9,9-dimethylfluorene

CAS Number: 108714-73-4
MDL Number: MFCD09953790
Molecular Formula: C15H15N
Molecular Weight: 209.29
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 168

Formula: C₁₅H₁₅N
Melting Pt: 168 °C
MDL Number: MFCD09953790
CAS Number: 108714-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df2d49a936ff6947bedabe892c4661d5e5c94ddf&vid=df2d49a936ff6947bedabe892c4661d5e5c94ddf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR