(Acetylthio)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-(Acetylthio)acetic acid

A2621-25G A2621-5G
TCA2621-25GEA 652.54 CAD
TCA2621-25G TCA2621-5G
(Acetylthio)acetic acid ≥98.0%
(Acetylthio)acetic acid

CAS Number: 1190-93-8
MDL Number: MFCD00021763
Molecular Formula: C4H6O3S
Molecular Weight: 134.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 160
Specific Gravity (20/20): 1.30

Formula: C₄H₆O₃S
MW: 134.15 g/mol
Boiling Pt: 160 °C
Density: 1.30 (20 °C)
MDL Number: MFCD00021763
CAS Number: 1190-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09593048c10613da204f50892130a49a4ce2c092&vid=09593048c10613da204f50892130a49a4ce2c092&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR