5-Amino-1-(2-hydroxyethyl)pyrazole ≥99.0% (by GC)

Supplier: TCI America

A2619-25G A2619-5G
TCA2619-25GEA 350.76 CAD
TCA2619-25G TCA2619-5G
5-Amino-1-(2-hydroxyethyl)pyrazole ≥99.0% (by GC)
5-Amino-1-(2-hydroxyethyl)pyrazole

CAS Number: 73616-27-0
MDL Number: MFCD08063809
Molecular Formula: C5H9N3O
Molecular Weight: 127.15
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 108

Formula: C₅H₉N₃O
Boiling Pt: 140 °C
Melting Pt: 108 °C
MDL Number: MFCD08063809
CAS Number: 73616-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6082edb18967d88c9ea3ec9c519058f56334ba0d&vid=6082edb18967d88c9ea3ec9c519058f56334ba0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR